4-Methyl-1H-pyrazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Fomepizole
AS22389G25
AS22389G5
ACTIAS22389G25EA
31
GBP
ACTIAS22389G25
ACTIAS22389G5
4-Methyl-1H-pyrazole
4-Methyl-1H-pyrazole
Formula:
C₄H₆N₂ MW: 82.11 g/mol |
MDL Number:
MFCD00005245 CAS Number: 7554-65-6 EINECS: 231-445-0 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...