2,3-Difluoro-6-nitroanisole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,2-Difluoro-3-methoxy-4-nitrobenzene
AS25233G100
AS25233G25
AS25233G5
ACTIAS25233G100EA
0
GBP
ACTIAS25233G100
ACTIAS25233G25
ACTIAS25233G5
2,3-Difluoro-6-nitroanisole
2,3-Difluoro-6-nitroanisole
Formula:
C₇H₅F₂NO₃ MW: 189.1163 g/mol |
MDL Number:
MFCD03840468 CAS Number: 66684-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...