2-Bromo-4-fluoro-6-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23345G1 AS23345G5
ACTIAS23345G1EA 0 GBP
ACTIAS23345G1 ACTIAS23345G5
2-Bromo-4-fluoro-6-nitrotoluene
2-Bromo-4-fluoro-6-nitrotoluene
Formula: C₇H₅BrFNO₂
MDL Number: MFCD03094189
CAS Number: 502496-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR