4-Chloro-6-methoxyquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2596G1 AS2596G5
ACTIAS2596G1EA 81 GBP
ACTIAS2596G1 ACTIAS2596G5
4-Chloro-6-methoxyquinazoline
4-Chloro-6-methoxyquinazoline
Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
MDL Number: MFCD06657615
CAS Number: 50424-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR