2,4-Dichloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20729G100 AS20729G25 AS20729G5
ACTIAS20729G100EA 283 GBP
ACTIAS20729G100 ACTIAS20729G25 ACTIAS20729G5
2,4-Dichloroquinazoline
2,4-Dichloroquinazoline
Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
MDL Number: MFCD00091950
CAS Number: 607-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR