6-Phenoxynicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21233G1 AS21233G25 AS21233G5
ACTIAS21233G1EA 0 GBP
ACTIAS21233G1 ACTIAS21233G25 ACTIAS21233G5
6-Phenoxynicotinic acid
6-Phenoxynicotinic acid
Formula: C₁₂H₉NO₃
MDL Number: MFCD02682068
CAS Number: 51362-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR