Cyclopropanesulphonamide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21566G100 AS21566G25 AS21566G5
ACTIAS21566G100EA 146 GBP
ACTIAS21566G100 ACTIAS21566G25 ACTIAS21566G5
Cyclopropanesulphonamide
Cyclopropanesulphonamide
Formula: C₃H₇NO₂S
MW: 121.15 g/mol
MDL Number: MFCD08705286
CAS Number: 154350-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR