3,5-Bis(trifluoromethyl)phenylhydrazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24983G25
AS24983G5
ACTIAS24983G25EA
0
GBP
ACTIAS24983G25
ACTIAS24983G5
3,5-Bis(trifluoromethyl)phenylhydrazine
3,5-Bis(trifluoromethyl)phenylhydrazine
Formula:
C₈H₆F₆N₂ MW: 244.14 g/mol |
MDL Number:
MFCD00051606 CAS Number: 886-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...