4-Bromo-2,6-dinitrotoluene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23910G1
AS23910G5
ACTIAS23910G1EA
67
GBP
ACTIAS23910G1
ACTIAS23910G5
4-Bromo-2,6-dinitrotoluene
4-Bromo-2,6-dinitrotoluene
Formula:
C₇H₅BrN₂O₄ MW: 261.03 g/mol |
MDL Number:
MFCD07357280 CAS Number: 95192-64-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...