2-Amino-4-(4-fluorophenyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23834G25 AS23834G5
ACTIAS23834G25EA 0 GBP
ACTIAS23834G25 ACTIAS23834G5
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
Formula: C₉H₇FN₂S
MW: 194.2287 g/mol
MDL Number: MFCD01086598
CAS Number: 77815-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR