Methyl-3-amino-5-hydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-amino-5-hydroxybenzoate

AS20305G1 AS20305G25 AS20305G5
ACTIAS20305G1EA 79 GBP
ACTIAS20305G1 ACTIAS20305G25 ACTIAS20305G5
Methyl-3-amino-5-hydroxybenzoate
Methyl 3-amino-5-hydroxybenzoate
Formula: C₈H₉NO₃
MW: 167.1620 g/mol
MDL Number: MFCD09263220
CAS Number: 67973-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR