4-Cyanophenylhydrazine hydrochloride
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydrazinylbenzonitrile hydrochloride
AS22976G100
AS22976G25
AS22976G5
ACTIAS22976G100EA
0
GBP
ACTIAS22976G100
ACTIAS22976G25
ACTIAS22976G5
4-Cyanophenylhydrazine hydrochloride
4-Cyanophenylhydrazine hydrochloride
Formula:
C₇H₈ClN₃ MW: 169.61 g/mol |
MDL Number:
MFCD00673994 CAS Number: 2863-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...