Methyl-2-formylbenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-formylbenzoate

AS35207G1 AS35207G5
ACTIAS35207G1EA 0 GBP
ACTIAS35207G1 ACTIAS35207G5
Methyl-2-formylbenzoate
Methyl 2-formylbenzoate
Formula: C₉H₈O₃
MDL Number: MFCD00204231
CAS Number: 4122-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR