3-Amino-3-(4-fluorophenyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22676G25 AS22676G5
ACTIAS22676G25EA 0 GBP
ACTIAS22676G25 ACTIAS22676G5
3-Amino-3-(4-fluorophenyl)propionic acid
3-Amino-3-(4-fluorophenyl)propionic acid
Formula: C₉H₁₀FNO₂
MDL Number: MFCD00181811
CAS Number: 325-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR