3-Amino-3-(2-fluorophenyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-3-(2-fluorophenyl)propanoic acid

AS24243G1 AS24243G25 AS24243G5
ACTIAS24243G1EA 39 GBP
ACTIAS24243G1 ACTIAS24243G25 ACTIAS24243G5
3-Amino-3-(2-fluorophenyl)propionic acid
3-Amino-3-(2-fluorophenyl)propionic acid
Formula: C₉H₁₀FNO₂
MW: 183.1796 g/mol
MDL Number: MFCD00193241
CAS Number: 117391-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR