L-Glutamic acid 5-methyl ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Glu(OMe)-OH

AS21048G100 AS21048G25
ACTIAS21048G100EA 0 GBP
ACTIAS21048G100 ACTIAS21048G25
L-Glutamic acid 5-methyl ester
L-Glutamic acid 5-methyl ester
Formula: C₆H₁₁NO₄
MW: 161.2 g/mol
MDL Number: MFCD00002632
CAS Number: 1499-55-4
EINECS: 216-110-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR