2,4-Dichloropyrimidine-5-carbonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dichloro-5-cyanopyrimidine

AS20893G1 AS20893G5
ACTIAS20893G1EA 21 GBP
ACTIAS20893G1 ACTIAS20893G5
2,4-Dichloropyrimidine-5-carbonitrile
2,4-Dichloropyrimidine-5-carbonitrile
Formula: C₅HCl₂N₃
MW: 173.9875 g/mol
MDL Number: MFCD09746248
CAS Number: 3177-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR