1-Phenyl-1H-pyrazole-4-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS1622G1 AS1622G5
ACTIAS1622G1EA 0 GBP
ACTIAS1622G1 ACTIAS1622G5
1-Phenyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-1H-pyrazole-4-carboxylic acid
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00514518
CAS Number: 1134-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR