1H-Pyrazole-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Pyrazolecarboxylic acid

ACTIAS20565G25EA 31 GBP
ACTIAS20565G25 ACTIAS20565G100 ACTIAS20565G5
1H-Pyrazole-3-carboxylic acid
1H-Pyrazole-3-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
MDL Number: MFCD00077436
CAS Number: 1621-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR