2-Amino-4-(3-pyridyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24135G1 AS24135G5
ACTIAS24135G1EA 0 GBP
ACTIAS24135G1 ACTIAS24135G5
2-Amino-4-(3-pyridyl)thiazole
2-Amino-4-(3-pyridyl)thiazole
Formula: C₈H₇N₃S
MDL Number: MFCD00218013
CAS Number: 30235-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR