4,5,7-Trichloro-2-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007452-1G 007452-5G
FLUO007452-1GEA 0 GBP
FLUO007452-1G FLUO007452-5G
4,5,7-Trichloro-2-(trifluoromethyl)quinoline
4,5,7-Trichloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₃Cl₃F₃N
MDL Number: MFCD00153279
CAS Number: 175203-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR