3,4-Dichlorobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013511-1G 013511-5G
FLUO013511-1GEA 0 GBP
FLUO013511-1G FLUO013511-5G
3,4-Dichlorobenzyl mercaptan
3,4-Dichlorobenzyl mercaptan
Formula: C₇H₆Cl₂S
MDL Number: MFCD00039651
CAS Number: 36480-40-7
EINECS: 253-054-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR