5-Bromo-2-methoxypyrimidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
016200-1G
016200-5G
FLUO016200-1GEA
0
GBP
FLUO016200-1G
FLUO016200-5G
5-Bromo-2-methoxypyrimidine
5-Bromo-2-methoxypyrimidine
Formula:
C₅H₅BrN₂O MW: 189.01 g/mol |
MDL Number:
MFCD04973351 CAS Number: 14001-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...