6-Fluoropicolinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid

005466-1G 005466-5G
FLUO005466-1GEA 0 GBP
FLUO005466-1G FLUO005466-5G
6-Fluoropicolinic acid
6-Fluoropicolinic acid
Formula: C₆H₄FNO₂
MDL Number: MFCD02181193
CAS Number: 402-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR