1-Boc-4-hydroxypiperidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008952-1G 008952-5G 008952-25G
FLUO008952-1GEA 0 GBP
FLUO008952-1G FLUO008952-5G FLUO008952-25G
1-Boc-4-hydroxypiperidine
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR