1-Benzhydrylpiperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019233-5G 019233-100G
FLUO019233-5GEA 0 GBP
FLUO019233-5G FLUO019233-100G
1-Benzhydrylpiperazine
1-Benzhydrylpiperazine
Formula: C₁₇H₂₀N₂
MDL Number: MFCD00038379
CAS Number: 841-77-0
EINECS: 212-667-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR