2,5-Dichloro-p-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

012740-100G 012740-25G
FLUO012740-100GEA 0 GBP
FLUO012740-100G FLUO012740-25G
2,5-Dichloro-p-xylene
2,5-Dichloro-p-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 222
Melting Pt: 71
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00000610
CAS Number: 1124-05-6
EINECS: 214-387-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR