2-Amino-5-pyridineboronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021607-5G 021607-1G
FLUO021607-5GEA 0 GBP
FLUO021607-5G FLUO021607-1G
2-Amino-5-pyridineboronic acid pinacol ester
2-Amino-5-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₇BN₂O₂
MDL Number: MFCD05663837
CAS Number: 827614-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR