1-Boc-5-fluoroindole-2-boronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Boc-5-fluoroindole-2-boronic Acid, N-(Boc)-5-fluoroindole-2-boronic acid
016247-1G
016247-5G
FLUO016247-1GEA
0
GBP
FLUO016247-1G
FLUO016247-5G
1-Boc-5-fluoroindole-2-boronic acid
1-Boc-5-fluoroindole-2-boronic acid
Formula:
C₁₃H₁₅BFNO₄ |
MDL Number:
MFCD04114581 CAS Number: 352359-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...