1H,1H,9H,9H-Perfluoro-1,9-nonanediol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H,1H,9H,9H-Perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonandiol
002306-5G
002306-25G
FLUO002306-5GEA
0
GBP
FLUO002306-5G
FLUO002306-25G
1H,1H,9H,9H-Perfluoro-1,9-nonanediol
1H,1H,9H,9H-Tetradecafluoro-1,9-nonandiol
Formula:
C₉H₆F₁₄O₂ |
MDL Number:
MFCD00153275 CAS Number: 203303-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...