1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol

002308-5G 002308-25G 002308-1G
FLUO002308-5GEA 0 GBP
FLUO002308-5G FLUO002308-25G FLUO002308-1G
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formula: C₁₂H₆F₂₀O₂
MW: 562.15 g/mol
MDL Number: MFCD00236625
CAS Number: 183162-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR