3'-Fluoro-5'-(trifluoromethyl)propiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006452-1G 006452-5G
FLUO006452-1GEA 0 GBP
FLUO006452-1G FLUO006452-5G
3'-Fluoro-5'-(trifluoromethyl)propiophenone
3'-Fluoro-5'-(trifluoromethyl)propiophenone
Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
MDL Number: MFCD00061262
CAS Number: 207974-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR