3-Fluoro-4-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Cyano-2-fluorobenzotrifluoride, 3-Fluoro-4-trifluoromethylbenzonitrile

007913-5G 007913-25G
FLUO007913-5GEA 0 GBP
FLUO007913-5G FLUO007913-25G
3-Fluoro-4-(trifluoromethyl)benzonitrile
3-Fluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 45…48 °C
MDL Number: MFCD00236280
CAS Number: 231953-38-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR