4-Bromo-3,5-xylenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013945-100G 013945-25G
FLUO013945-100GEA 0 GBP
FLUO013945-100G FLUO013945-25G
4-Bromo-3,5-xylenol
4-Bromo-3,5-xylenol
Formula: C₈H₉BrO
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 116 °C
MDL Number: MFCD00002315
CAS Number: 7463-51-6
EINECS: 231-255-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR