3-Fluoro-5-(trifluoromethyl)benzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006426-1G
006426-5G
FLUO006426-1GEA
0
GBP
FLUO006426-1G
FLUO006426-5G
3-Fluoro-5-(trifluoromethyl)benzyl bromide
3-Fluoro-5-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrF₄ Boiling Pt: 43 °C (1 mmHg) |
MDL Number:
MFCD00061175 CAS Number: 239087-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...