4-Fluoro-2-(trifluoromethyl)benzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006420-1G 006420-5G
FLUO006420-1GEA 0 GBP
FLUO006420-1G FLUO006420-5G
4-Fluoro-2-(trifluoromethyl)benzylamine
4-Fluoro-2-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 46 °C
MDL Number: MFCD00061164
CAS Number: 202522-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR