2-Fluoro-5-(trifluoromethyl)benzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006417-1G
006417-5G
FLUO006417-1GEA
0
GBP
FLUO006417-1G
FLUO006417-5G
2-Fluoro-5-(trifluoromethyl)benzylamine
2-Fluoro-5-(trifluoromethyl)benzylamine
Formula:
C₈H₇F₄N Boiling Pt: 185 °C (1013 hPa) Melting Pt: 46 °C |
MDL Number:
MFCD00061162 CAS Number: 199296-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...