2-Fluoro-3-(trifluoromethyl)benzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007921-1G 007921-5G
FLUO007921-1GEA 0 GBP
FLUO007921-1G FLUO007921-5G
2-Fluoro-3-(trifluoromethyl)benzylamine
2-Fluoro-3-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
Boiling Pt: 193 °C (1013 hPa)
Density: 1.321 g/cm³ (20 °C)
MDL Number: MFCD00061161
CAS Number: 239135-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR