4-Nitro-3-picoline-N-oxide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide

OS-7541-5G OS-7541-25G OS-7541-100G
COBBOS-7541-5GEA 0 GBP
COBBOS-7541-5G COBBOS-7541-25G COBBOS-7541-100G
4-Nitro-3-picoline-N-oxide
4-Nitro-3-picoline N-oxide
Formula: C₆H₆N₂O₃
Melting Pt: 137.5 °C
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR