2-Fluoro-5-(trifluoromethyl)benzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006404-1G
006404-5G
006404-10G
FLUO006404-1GEA
0
GBP
FLUO006404-1G
FLUO006404-5G
FLUO006404-10G
2-Fluoro-5-(trifluoromethyl)benzoyl chloride
2-Fluoro-5-(trifluoromethyl)benzoyl chloride
Formula:
C₈H₃ClF₄O MW: 226.56 g/mol Boiling Pt: 190…191 °C (1013 hPa) Density: 1.499 g/cm³ (20 °C) |
MDL Number:
MFCD00061156 CAS Number: 207981-46-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...