7-Methoxyindole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
012717-1G
012717-5G
FLUO012717-1GEA
0
GBP
FLUO012717-1G
FLUO012717-5G
7-Methoxyindole
7-Methoxyindole
Formula:
C₉H₉NO Boiling Pt: 298 °C (1013 hPa) Density: 1,126 g/cm³ (25 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00047203 CAS Number: 3189-22-8 EINECS: 221-690-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...