3-Bromo-2-fluoropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
016228-1G
016228-5G
FLUO016228-1GEA
0
GBP
FLUO016228-1G
FLUO016228-5G
3-Bromo-2-fluoropyridine
3-Bromo-2-fluoropyridine
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04112496 CAS Number: 36178-05-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...