1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Perfluoropropoxyethylene, (Heptafluoropropoxy)trifluoroethene, Perfluoro(propyl vnyl ether)

010522-25G 010522-5G
FLUO010522-25GEA 0 GBP
FLUO010522-25G FLUO010522-5G
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane
Formula: C₅F₁₀O
MDL Number: MFCD00191577
CAS Number: 1623-05-8
EINECS: 216-600-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR