4-Hydroxyindole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
016235-1G
016235-25G
016235-5G
FLUO016235-1GEA
0
GBP
FLUO016235-1G
FLUO016235-25G
FLUO016235-5G
4-Hydroxyindole
4-Hydroxyindole
Formula:
C₈H₇NO Boiling Pt: 100 °C (0.01 mmHg) Melting Pt: 102…104 °C |
MDL Number:
MFCD00005667 CAS Number: 2380-94-1 EINECS: 219-177-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...