(1R,2R)-(-)-1,2-Diaminocyclohexane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

012705-1G 012705-5G
FLUO012705-1GEA 0 GBP
FLUO012705-1G FLUO012705-5G
(1R,2R)-(-)-1,2-Diaminocyclohexane
(1R,2R)-(-)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 14…15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR