4'-Cyano-2'-(trifluoromethyl)acetanilide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004594-1G
004594-5G
FLUO004594-1GEA
0
GBP
FLUO004594-1G
FLUO004594-5G
4'-Cyano-2'-(trifluoromethyl)acetanilide
4'-Cyano-2'-(trifluoromethyl)acetanilide
Formula:
C₁₀H₇F₃N₂O |
MDL Number:
MFCD00052844 CAS Number: 175277-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...