3-Methyl-2-thiophenecarbonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1303-25G HI-1303-5G
COBBHI-1303-25GEA 0 GBP
COBBHI-1303-25G COBBHI-1303-5G
3-Methyl-2-thiophenecarbonitrile
3-Methyl-2-thiophenecarbonitrile
Formula: C₆H₅NS
MDL Number: MFCD00052086
CAS Number: 55406-13-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR