5-Bromo-4-cyclohexylpyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6081-5G HC-6081-1G
COBBHC-6081-5GEA 0 GBP
COBBHC-6081-5G COBBHC-6081-1G
5-Bromo-4-cyclohexylpyrimidine
5-Bromo-4-cyclohexylpyrimidine
Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD09475877
CAS Number: 941294-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR