Bis(2-methoxyethyl)amine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B48207-1L B48207-250ML
SIALB48207-1LEA 778 GBP
SIALB48207-1L SIALB48207-250ML
Bis(2-methoxyethyl)amine, Sigma-Aldrich®
Bis(2-methoxyethyl)amine
Formula: C₆H₁₅NO₂
MDL Number: MFCD00025906
CAS Number: 111-95-5
EINECS: 203-923-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR