N-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF673.5 008407-5G 008407-25G
CHMPCFF673.5EA 0 GBP
CHMPCFF673.5 FLUO008407-5G FLUO008407-25G
N-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine
N-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine
Formula: C₁₁H₁₁F₃N₂O₂
MDL Number: MFCD00052886
CAS Number: 40832-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR